Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl alcohol, Pure, 64-17-5, MFCD00003568, 4L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 4598284L
Encompass
Linear Formula CH3CH2OH, ≥99.5%, Molecular Weight 46.07, 200 proof, HPLC/spectrophotometric grade, Synonym: Ethanol, absolute alcohol, non-denatured ethanol
Catalog No. 50-165-7087