Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol Standard, 10% (v/v), Ricca Chemical
SDP

Catalog No. R291910050A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R291910050A 50 mL Natural Poly Bottle
R2919100500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R291910050A Supplier Ricca Chemical Company Supplier No. R291910050A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Packaging Natural Poly Bottle
CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Melting Point Approximately 0°C
Type Standard
Molecular Weight (g/mol) 46.069
Quantity 50 mL
Concentration or Composition (by Analyte or Components) 10% (v/v)
Chemical Name or Material Ethyl Alcohol Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.