Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl all cis-5,8,11,14,17-Eicosapentaenoate (stabilized with Tocopherols) 65.0+%, TCI America™
SDP

Catalog No. E085325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E085325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E085325G Supplier TCI America Supplier No. E085325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 86227-47-6
Molecular Formula C22H34O2
Molecular Weight (g/mol) 330.512
MDL Number MFCD00673476
InChI Key SSQPWTVBQMWLSZ-AAQCHOMXSA-N
Synonym all cis-5,8,11,14,17-Eicosapentaenoic Acid Ethyl Ester, EPA Ethyl Ester, Ethyl all cis-5,8,11,14,17-Icosapentaenoate
PubChem CID 9831415
ChEBI CHEBI:84883
IUPAC Name ethyl (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoate
SMILES CCC=CCC=CCC=CCC=CCC=CCCCC(=O)OCC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 330.51
Percent Purity ≥65.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl all cis-5,8,11,14,17-Eicosapentaenoate (stabilized with Tocopherols)
RTECSNumber JX3840000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.