Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Allophanate 98.0+%, TCI America™
SDP

Catalog No. A021325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A021325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A021325G Supplier TCI America Supplier No. A021325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent used in chemical research and synthesis applications.

Chemical Identifiers

CAS 626-36-8
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD00047870
InChI Key PIHPSKJRLDSJPX-UHFFFAOYSA-N
Synonym Allophanic Acid Ethyl Ester
PubChem CID 69380
IUPAC Name ethyl N-carbamoylcarbamate
SMILES CCOC(=O)NC(N)=O

Specifications

Melting Point 190°C
Color White
Quantity 25 g
Formula Weight 132.12
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Allophanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.