Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl alpha-bromoisobutyrate

Catalog No. E14403100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E14403100G 100 g
E144035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E14403100G Supplier Sigma Aldrich Supplier No. E14403100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Atom Transfer Radical Polymerization (ATRP) initiator that will generate a polymer with an ethyl end group.

Specifications

CAS 600-00-0
Density 1.329 g/mL (at 25°C (literature))
Boiling Point 65°C to 67°C (11 mmHg)
Molecular Formula C6H11BrO2
Refractive Index n20/D 1.444 (literature)
Linear Formula (CH3)2 CBrCOOC2H5
MDL Number MFCD00000123
Quantity 100 g
Synonym Ethyl 2-bromo-2-methylpropionate
Molecular Weight (g/mol) 195.05
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.