Learn More
Medchemexpress LLC Ethyl arachidonate | 1808-26-0 | 98.32% | 332.52 | 1 ML
SDP
Catalog No. 5000454155
Encompass_Preferred
Ethyl arachidonate is a lipophilic esterified form of arachidonic acid (AA). It can be incorporated into dietary regimens or supplied to cultured cells as a source of exogenous arachidonate. It is the primary fatty acid ethyl ester (FAEE) found in the brains of individuals who have been intoxicated with alcohol.
  • Source of exogenous arachidonate
  • Main species of fatty acid ethyl esters (FAEE) in brain of alcohol-intoxicated subjects
  • Colorless to light yellow liquid
Catalog No. 50-004-54155 Supplier Medchemexpress LLC Supplier No. HYW01321410MM/1ML
Ethyl arachidonate is a lipophilic esterified form of arachidonic acid (AA). It can be incorporated into dietary regimens or supplied to cultured cells as a source of exogenous arachidonate. It is the primary fatty acid ethyl ester (FAEE) found in the brains of individuals who have been intoxicated with alcohol.
  • Source of exogenous arachidonate
  • Main species of fatty acid ethyl esters (FAEE) in brain of alcohol-intoxicated subjects
  • Colorless to light yellow liquid