Learn More
Medchemexpress LLC Ethyl arachidonate | 1808-26-0 | 98.32% | 332.52 | 25 MG
SDP
Catalog No. 5000366679
Encompass_Preferred
Ethyl arachidonate is a lipophilic esterified form of arachidonic acid (AA) that can be incorporated into dietary regimens or fed to cultured cells as an exogenous arachidonate source. It is also identified as the primary species of fatty acid ethyl esters (FAEE) found in the brains of individuals who have been alcohol-intoxicated.
  • Lipophilic esterified form of arachidonic acid
  • Can be incorporated into dietary regimens
  • Can be fed to cultured cells as an exogenous arachidonate source
  • Primary species of fatty acid ethyl esters (FAEE) in alcohol-intoxicated brains
Catalog No. 50-003-66679 Supplier Medchemexpress LLC Supplier No. 25MGHYW013214
Ethyl arachidonate is a lipophilic esterified form of arachidonic acid (AA) that can be incorporated into dietary regimens or fed to cultured cells as an exogenous arachidonate source. It is also identified as the primary species of fatty acid ethyl esters (FAEE) found in the brains of individuals who have been alcohol-intoxicated.
  • Lipophilic esterified form of arachidonic acid
  • Can be incorporated into dietary regimens
  • Can be fed to cultured cells as an exogenous arachidonate source
  • Primary species of fatty acid ethyl esters (FAEE) in alcohol-intoxicated brains