Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl azetidine-3-carboxylate hydrochloride | 405090-31-5 | 98.0% | 165.62 | C6H12ClNO2 | 10 G
SDP

Catalog No. 5000202945
Encompass_Preferred

Ethyl azetidine-3-carboxylate hydrochloride is the hydrochloride salt of ethyl azetidine-3-carboxylate supplied as a research reagent and ADC linker intermediate. It is intended for medicinal chemistry applications and antibody-drug conjugate linker construction. Purity is 98.0% and molecular weight is 165.62 g/mol, with standardized safety and quality documents available.

  • Hydrochloride salt of ethyl azetidine-3-carboxylate.
  • Purity 98.0%.
  • Molecular weight 165.62 g/mol.
  • CAS number 405090-31-5.
  • Available in 5 g, 10 g, 25 g, and 50 g pack sizes.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-002-02945 Supplier Medchemexpress LLC Supplier No. HYW05260010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl azetidine-3-carboxylate hydrochloride is the hydrochloride salt of ethyl azetidine-3-carboxylate supplied as a research reagent and ADC linker intermediate. It is intended for medicinal chemistry applications and antibody-drug conjugate linker construction. Purity is 98.0% and molecular weight is 165.62 g/mol, with standardized safety and quality documents available.

  • Hydrochloride salt of ethyl azetidine-3-carboxylate.
  • Purity 98.0%.
  • Molecular weight 165.62 g/mol.
  • CAS number 405090-31-5.
  • Available in 5 g, 10 g, 25 g, and 50 g pack sizes.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.