Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl azetidine-3-carboxylate hydrochloride | 405090-31-5 | 98.0% | 165.62 g/mol | C6H12ClNO2 | 5 G
SDP

Catalog No. 5000202929
Encompass_Preferred

Ethyl azetidine-3-carboxylate hydrochloride is the hydrochloride salt of ethyl azetidine-3-carboxylate, used as a synthetic building block in medicinal chemistry and antibody-drug conjugate linker synthesis.

  • Hydrochloride salt supplied as a powder.
  • Purity 98.0% (NMR).
  • Molecular formula C6H12ClNO2 and molecular weight 165.62 g/mol.
  • Available in laboratory-scale quantities, including 5 g packs.
  • Recommended storage -20°C per certificate of analysis.

Catalog No. 50-002-02929 Supplier Medchemexpress LLC Supplier No. HYW0526005G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl azetidine-3-carboxylate hydrochloride is the hydrochloride salt of ethyl azetidine-3-carboxylate, used as a synthetic building block in medicinal chemistry and antibody-drug conjugate linker synthesis.

  • Hydrochloride salt supplied as a powder.
  • Purity 98.0% (NMR).
  • Molecular formula C6H12ClNO2 and molecular weight 165.62 g/mol.
  • Available in laboratory-scale quantities, including 5 g packs.
  • Recommended storage -20°C per certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.