Learn More
AdipoGen Ethyl azidoacetate Solution, 25% in toluene (25 ml)
SDP
Supplier:  AdipoGen CDXE0531L025
Encompass_Preferred
Chemical. CAS: 637-81-0. Formula: C4H7N3O2. MW: 129.12. Synthetic. Building block for synthesis. Useful precursors to N-heterocycles.
Catalog No. 50-205-4429