Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl azidoacetate, 25% solution in toluene

Catalog No. AC448780050
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448780050 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC448780050 Supplier Thermo Scientific Chemicals Supplier No. 448780050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 637-81-0 | C4H7N3O2 | 129.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 637-81-0, 108-88-3
Density 0.9200g/mL
Flash Point 7°C
Packaging Glass bottle
Molecular Formula C4H7N3O2
Quantity 5 mL
Specific Gravity 0.92
Synonym ethyl azidoacetate, azidoacetic acid ethyl ester, acetic acid, azido-, ethyl ester, azido-acetic acid ethyl ester, ccris 3368, acetic acid, 2-azido-,ethyl ester, ethyl azido-acetate, ethyl2-azidoacetate, ethyl-2-azidoacetate, ethylazidoacetatesolution
InChI Key HVJJYOAPXBPQQV-UHFFFAOYSA-N
SMILES CCOC(=O)CN=[N+]=[N-]
IUPAC Name ethyl 2-azidoacetate
Molecular Weight (g/mol) 129.12
PubChem CID 69480
Formula Weight 129.12
Chemical Name or Material Ethyl azidoacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Suspected of damaging the unborn child.
May be fatal if swallowed and
EINECSNumber 211-301-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.