Learn More
Medchemexpress LLC Ethyl benzoate | 93-89-0 | MFCD00009109 | 99.9% | 150.17 g/mol | C9H10O2 | 1 ML
SDP
Catalog No. 5000301380
Encompass_Preferred
Ethyl benzoate is an organic ester (C9H10O2) provided as a ready-to-use 10 mM solution in DMSO for research and analytical applications. It is used as a reference standard and reagent for analytical method development, chromatography, and biochemical assays that require a defined concentration and high purity (CAS 93-89-0).
  • Ready-to-use 10 mM solution in DMSO.
  • High purity suitable for analytical use.
  • Convenient 1 mL volume for small-scale assays.
  • Accompanied by safety data and certificate of analysis.
  • Suitable for chromatography and method development as a reference standard.
Catalog No. 50-003-01380 Supplier Medchemexpress LLC Supplier No. HYW01743910MM/1ML
Ethyl benzoate is an organic ester (C9H10O2) provided as a ready-to-use 10 mM solution in DMSO for research and analytical applications. It is used as a reference standard and reagent for analytical method development, chromatography, and biochemical assays that require a defined concentration and high purity (CAS 93-89-0).
  • Ready-to-use 10 mM solution in DMSO.
  • High purity suitable for analytical use.
  • Convenient 1 mL volume for small-scale assays.
  • Accompanied by safety data and certificate of analysis.
  • Suitable for chromatography and method development as a reference standard.