Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl benzoate | 93-89-0 | MFCD00009109 | 99.9% | 150.17 g/mol | C9H10O2 | 100 G
SDP

Catalog No. 5000301410
Encompass_Preferred

Ethyl benzoate is the ethyl ester of benzoic acid supplied as an analytical reference standard for research and analytical applications. It is provided as a colorless to light-yellow liquid and is used as a reference material in chromatography and analytical testing. The material is intended for laboratory research use only.

  • High purity (99.94%).
  • Liquid, colorless to light yellow.
  • Molecular formula C9H10O2.
  • Molecular weight 150.17 g/mol.
  • CAS number 93-89-0.
  • Stable under recommended storage conditions.
  • Intended for research use only; not for human use.

Catalog No. 50-003-01410 Supplier Medchemexpress LLC Supplier No. HYW017439100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl benzoate is the ethyl ester of benzoic acid supplied as an analytical reference standard for research and analytical applications. It is provided as a colorless to light-yellow liquid and is used as a reference material in chromatography and analytical testing. The material is intended for laboratory research use only.

  • High purity (99.94%).
  • Liquid, colorless to light yellow.
  • Molecular formula C9H10O2.
  • Molecular weight 150.17 g/mol.
  • CAS number 93-89-0.
  • Stable under recommended storage conditions.
  • Intended for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.