Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl benzoylformate

Catalog No. 2589115G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-891-15G 5 g
25-891-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-891-15G Supplier Sigma Aldrich Supplier No. 2589115G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Enantioselective hydrogenation of ethyl benzoylformate on Pt/Al2O3 modified with dihydrocinchonidine has been studied.Ethyl benzoylformate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.

Specifications

CAS 1603-79-8
Density 1.122 g/mL (at 25°C (literature))
Boiling Point 138°C to 139°C (18 mmHg)
Molecular Formula C10H10O3
Refractive Index n20/D 1.516 (literature)
Linear Formula C6H5COCO2C2H5
MDL Number MFCD00009120
Quantity 5 g
Synonym Ethyl phenylglyoxylate
Molecular Weight (g/mol) 178.18
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.