Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl bromoacetate

Catalog No. 17020500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-020-500ML 500 mL
17-020-100ML 100 mL
17-020-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-020-500ML Supplier Sigma Aldrich Supplier No. 17020500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl bromoacetate was used in the synthesis of:
  • Artificial diethylstilbestrol antigen
  • 3-phenyl-1-naphthol, a key intermediate in the synthesis of vanol
  • Novel pyrazolothiazol-4(5h)-ones
  • Steroidal thiazolidinone derivatives

Specifications

CAS 105-36-2
Density 1.506 g/mL (at 25°C (literature))
Boiling Point 159°C (lit.)
Molecular Formula C4H7BrO2
Refractive Index n20/D 1.451 (literature); n20/D 1.451
Linear Formula BrCH2COOC2H5
MDL Number MFCD00000191
Quantity 500 mL
Molecular Weight (g/mol) 167
Percent Purity ≥97.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.