Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl bromoacetate

Catalog No. 133973100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-397-3100G 100 g
13-397-3500G 500 g
13-397-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-397-3100G Supplier Sigma Aldrich Supplier No. 133973100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl bromoacetate on derivatisation reaction with p-t-butyl calix[4]arene yields 1,3-diester substituted calix[4]arene
  • It also undergoes suzuki type cross-coupling reactions with arylboronic acids cocatalyzed by copper(i) oxide.ethyl bromoacetate was used to prepare reversibly photoresponsive coumarin-stabilized polymeric nanoparticles in an aqueous medium, which act as a detectable drug carrier

Specifications

CAS 105-36-2
Density 1.506 g/mL (at 25°C (literature))
Boiling Point 159°C (lit.)
Molecular Formula C4H7BrO2
Refractive Index n20/D 1.451 (literature)
Linear Formula BrCH2COOC2H5
MDL Number MFCD00000191
Quantity 100 g
Molecular Weight (g/mol) 167
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.