Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl butyrate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019342 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9342 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9342 Supplier MilliporeSigma Supelco Supplier No. 755635ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl butyrate belongs to the class of esters and is generally employed as a flavor enhancer in commercial food products.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 865°F
CAS 105-54-4
Vapor Pressure 15.5 mmHg (25°C)
Vapor Density 4 (vs. air)
Density 0.875 g/mL (at 25°C)
Flash Point 26°C
Physical Form Neat
Linear Formula CH3 CH2 CH2 C(O)OC2H5
MDL Number MFCD00009394
Melting Point -93°C
Boiling Point 120°C
Synonym Butyric acid ethyl ester
Type Analytical Standard
Quantity 5 mL
Formula Weight 116.16
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.