Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl butyrylacetate | 3249-68-1 | MFCD00009401 | 96.0% | 158.20 g/mol | C8H14O3 | 500 G
SDP

Catalog No. 5000302958
Encompass_Preferred

Ethyl butyrylacetate (Ethyl 3-oxohexanoate, CAS 3249-68-1) is an organic ester with a fruity aroma used as a flavor and fragrance intermediate, reagent, and research chemical. Supplied as a colorless to light-yellow liquid, the material is intended for laboratory and formulation use and is for research use only.

  • Purity around 96.0% as specified on the product record.
  • Molecular weight 158.20 g/mol and formula C8H14O3.
  • Colorless to light-yellow liquid, density ~0.989 g/cm³.
  • Suitable as a flavor and fragrance intermediate and synthetic reagent.
  • Available in multiple pack sizes, including 500 g, for bench-scale work.
  • Stable under recommended storage: -20°C (3 years) or 4°C (2 years) in pure form.

Catalog No. 50-003-02958 Supplier Medchemexpress LLC Supplier No. HYW015487500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl butyrylacetate (Ethyl 3-oxohexanoate, CAS 3249-68-1) is an organic ester with a fruity aroma used as a flavor and fragrance intermediate, reagent, and research chemical. Supplied as a colorless to light-yellow liquid, the material is intended for laboratory and formulation use and is for research use only.

  • Purity around 96.0% as specified on the product record.
  • Molecular weight 158.20 g/mol and formula C8H14O3.
  • Colorless to light-yellow liquid, density ~0.989 g/cm³.
  • Suitable as a flavor and fragrance intermediate and synthetic reagent.
  • Available in multiple pack sizes, including 500 g, for bench-scale work.
  • Stable under recommended storage: -20°C (3 years) or 4°C (2 years) in pure form.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.