Learn More
Medchemexpress LLC Ethyl Caffeate | 102-37-4 | 100 MG
SDP
Catalog No. 5000417989
Encompass_Preferred
Ethyl Caffeate is a natural phenolic compound isolated from *Bidens pilosa*. It suppresses NF-κB activation and its downstream inflammatory mediators, including inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), and prostaglandin E2 (PGE2) in vitro and in mouse skin. This product is for research use only.
  • Natural phenolic compound
  • Suppresses NF-κB activation
  • Targets inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), and prostaglandin E2 (PGE2)
  • For research use only
Catalog No. 50-004-17989 Supplier Medchemexpress LLC Supplier No. HYN6966100MG
Ethyl Caffeate is a natural phenolic compound isolated from *Bidens pilosa*. It suppresses NF-κB activation and its downstream inflammatory mediators, including inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), and prostaglandin E2 (PGE2) in vitro and in mouse skin. This product is for research use only.
  • Natural phenolic compound
  • Suppresses NF-κB activation
  • Targets inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), and prostaglandin E2 (PGE2)
  • For research use only