Learn More
Medchemexpress LLC Ethyl Caffeate | 102-37-4 | 50 MG
SDP
Catalog No. 5000417234
Encompass_Preferred
Ethyl Caffeate is a natural phenolic compound isolated from Bidens pilosa. It functions as a COX Inhibitor and suppresses NF-κB activation and its downstream inflammatory mediators, such as inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), and prostaglandin E2 (PGE2), in both in vitro settings and mouse skin. It is intended for research use only and is not sold to patients.
  • Natural phenolic compound
  • Functions as a COX Inhibitor
  • Suppresses NF-κB activation and downstream inflammatory mediators
  • Purity of 99.34%
  • Appears as a light yellow to yellow solid
Catalog No. 50-004-17234 Supplier Medchemexpress LLC Supplier No. HYN696650MG
Ethyl Caffeate is a natural phenolic compound isolated from Bidens pilosa. It functions as a COX Inhibitor and suppresses NF-κB activation and its downstream inflammatory mediators, such as inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), and prostaglandin E2 (PGE2), in both in vitro settings and mouse skin. It is intended for research use only and is not sold to patients.
  • Natural phenolic compound
  • Functions as a COX Inhibitor
  • Suppresses NF-κB activation and downstream inflammatory mediators
  • Purity of 99.34%
  • Appears as a light yellow to yellow solid