Learn More
Medchemexpress LLC Ethyl Caffeate | 102-37-4 | MFCD00045754 | 1 ML
SDP
Catalog No. 5000417938
Encompass_Preferred
Ethyl Caffeate is a natural phenolic compound isolated from Bidens pilosa. It suppresses NF-κB activation and its downstream inflammatory mediators, inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), and prostaglandin E2 (PGE2) in vitro or in mouse skin. It is for research use only.
  • Isolated from Bidens pilosa
  • Suppresses NF-κB activation
  • Inhibits iNOS, COX-2, and PGE2
  • For research use only
Catalog No. 50-004-17938 Supplier Medchemexpress LLC Supplier No. HYN696610MM/1ML
Ethyl Caffeate is a natural phenolic compound isolated from Bidens pilosa. It suppresses NF-κB activation and its downstream inflammatory mediators, inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), and prostaglandin E2 (PGE2) in vitro or in mouse skin. It is for research use only.
  • Isolated from Bidens pilosa
  • Suppresses NF-κB activation
  • Inhibits iNOS, COX-2, and PGE2
  • For research use only