Learn More
Medchemexpress LLC Ethyl β-carboline-3-carboxylate | 74214-62-3 | MFCD00009730 | 99.8% | C14H12N2O2 | 10MG
SDP
Catalog No. 5000259653
Encompass_Preferred
Ethyl β-carboline-3-carboxylate is a β-carboline derivative that functions as a ligand and short-acting antagonist at benzodiazepine receptors. It is used in preclinical research to probe benzodiazepine receptor pharmacology and to study interactions with diazepam, including modulation of cerebellar cGMP levels.
  • Ligand and short-acting antagonist of benzodiazepine receptors.
  • Useful for preclinical studies of benzodiazepine pharmacology and receptor modulation.
  • Demonstrated interaction with diazepam affecting cerebellar cGMP levels.
  • High purity (99.75%) suitable for research applications.
  • Solid, off-white to light yellow appearance for easy handling.
  • Storage: sealed and away from moisture; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
Catalog No. 50-002-59653 Supplier Medchemexpress LLC Supplier No. HY13749410MG
Ethyl β-carboline-3-carboxylate is a β-carboline derivative that functions as a ligand and short-acting antagonist at benzodiazepine receptors. It is used in preclinical research to probe benzodiazepine receptor pharmacology and to study interactions with diazepam, including modulation of cerebellar cGMP levels.
  • Ligand and short-acting antagonist of benzodiazepine receptors.
  • Useful for preclinical studies of benzodiazepine pharmacology and receptor modulation.
  • Demonstrated interaction with diazepam affecting cerebellar cGMP levels.
  • High purity (99.75%) suitable for research applications.
  • Solid, off-white to light yellow appearance for easy handling.
  • Storage: sealed and away from moisture; in solvent store at -80°C up to 6 months or -20°C up to 1 month.