Learn More
Medchemexpress LLC Ethyl cellulose 1kg
SDP
Catalog No. 5000205060
Encompass_Preferred
Ethyl cellulose is a derivative of cellulose Ethyl cellulose is a non-toxic biodegradable polymer Ethyl cellulose has unique properties such as oil gel formation active ingredient delivery and film formation Ethyl cellulose can be used as a pharmaceutical excipient such as a coating agent flavoring agent tablet filler etc Ethyl cellulose can be used to prepare nanoparticles for active compound delivery[1][2][3][4]
Catalog No. 50-002-05060 Supplier Medchemexpress LLC Supplier No. HYW1157461KG
Ethyl cellulose is a derivative of cellulose Ethyl cellulose is a non-toxic biodegradable polymer Ethyl cellulose has unique properties such as oil gel formation active ingredient delivery and film formation Ethyl cellulose can be used as a pharmaceutical excipient such as a coating agent flavoring agent tablet filler etc Ethyl cellulose can be used to prepare nanoparticles for active compound delivery[1][2][3][4]