Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl Chlorooxoacetate

Catalog No. E4310125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E4310125G 25 g
E43101100G 100 g
E43101500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. E4310125G Supplier Sigma Aldrich Supplier No. E4310125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl chlorooxoacetate can be used for the synthesis of:+a-keto esters.Functionalized 3-pyrolin-2-ones.Substituted arylglyoxylic acids via Friedel-Crafts acylation.Substituted 9,10-phenanthrenequinones.Quinoxalinone derivatives.

Specifications

CAS 1530-32-1
Melting Point 203°C to 205°C
Molecular Formula C20H20BrP
Refractive Index n20/D 1.416
Linear Formula [(C6H5)3P+C2H5]Br-
MDL Number MFCD00011838
Quantity 25 g
Synonym mono-Ethyl oxalyl chloride; Ethyl chloroglyoxylate; Monoethyl oxalyl chloride; Oxalic acid monoethyl ester chloride
Molecular Weight (g/mol) 371.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.