Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl cinnamate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012342 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2342 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2342 Supplier MilliporeSigma Supelco Supplier No. 66761100MG

missing translation for 'validMainframeMsg'

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 103-36-6
Density 1.049 g/mL (at 20°C)
Physical Form Neat
Linear Formula C6H5 CH=CHCOOC2H5
MDL Number MFCD00009189
Melting Point 6°C to 8°C
Boiling Point 271°C
Synonym 3-Phenyl-2-propenoic acid ethyl ester; Ethyl 3-phenyl-2-propenoate; NSC 6773
Type Analytical Standard
Quantity 100 mg
Formula Weight 176.21
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.