Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl cyanoformate

Catalog No. E1885950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1885950G 50 g
E1885910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1885950G Supplier Sigma Aldrich Supplier No. E1885950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl cyanoformate is a general cyanating agent
  • It can be used in the nucleophilic addition of cyanide to carbonyl compounds, oxidative cyanation of tertiary amines and in the synthesis of α-cyano-substituted acrylates from alkynes

Specifications

CAS 623-49-4
Density 1.003 g/mL (at 25°C (literature))
Boiling Point 115°C to 116°C (lit.)
Molecular Formula C4H5NO2
Refractive Index n20/D 1.381 (literature)
Linear Formula NCCO2C2H5
MDL Number MFCD00001836
Quantity 50 g
Molecular Weight (g/mol) 99.09
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.