Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Cyclobutanecarboxylate 98.0+%, TCI America™
SDP

Catalog No. E09315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09315G 5 g
E093125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09315G Supplier TCI America Supplier No. E09315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14924-53-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001321
InChI Key SMVBADCAMQOTOV-UHFFFAOYSA-N
Synonym Cyclobutanecarboxylic Acid Ethyl Ester
PubChem CID 84700
IUPAC Name ethyl cyclobutanecarboxylate
SMILES CCOC(=O)C1CCC1

Specifications

Color Yellow
Boiling Point 159°C
Quantity 5 g
UN Number 3272
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Cyclobutanecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.