Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl cyclohexanecarboxylate, 98+%

Catalog No. AAL0262206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0262206 5 g
AAL0262214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0262206 Supplier Thermo Scientific Chemicals Supplier No. L0262206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3289-28-9 | C9H16O2 | 156.225 g/mol

Ethyl cyclohexanecarboxylate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclohexanecarboxylate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3289-28-9
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00041734
InChI Key JJOYCHKVKWDMEA-UHFFFAOYSA-N
Synonym cyclohexanecarboxylic acid, ethyl ester, ethyl cyclohexylformate, ethyl hexahydrobenzoate, cyclohexanecarboxylic acid ethyl ester, ethoxycarbonylcyclohexane, ethyl cyclohexylcarboxylate, ethyl cyclohexanoate, fema no. 3544, ethyl cyclohexylmethanoate, ethyl cyclohexanocarboxylate
PubChem CID 18686
IUPAC Name ethyl cyclohexanecarboxylate
SMILES CCOC(=O)C1CCCCC1

Specifications

Density 0.936
Boiling Point 194°C to 196°C
Refractive Index 1.442
Quantity 5 g
Beilstein 2042698
Solubility Information Insoluble in water.
Formula Weight 156.23
Percent Purity ≥98%
Chemical Name or Material Ethyl cyclohexanecarboxylate
EINECSNumber 221-945-7
RTECSNumber GU8490000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.