Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl cyclohexylideneacetate, 98%

Catalog No. AAB2507306
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1552-92-7 | C10H16O2 | 168.24 g/mol

Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.

Solubility
Soluble in dichloromethane, ethyl acetate.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1552-92-7
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00017627,MFCD00963857
InChI Key MCWDXHYYYNGYGK-UHFFFAOYSA-N
Synonym ethyl cyclohexylideneacetate, acetic acid, cyclohexylidene-, ethyl ester, ethyl 2-cyclohexylidenacetate, cyclohexylideneacetic acid ethyl ester, ethoxycarbonylmethylenecyclohexane, .delta.1,.alpha.-cyclohexaneacetic acid, ethyl ester, acmc-20ahxr, ethylcyclohexylidenacetate, .delta.1, ethyl ester, ethyl2-cyclohexylideneacetate
PubChem CID 73776
IUPAC Name ethyl 2-cyclohexylideneacetate
SMILES CCOC(=O)C=C1CCCCC1

Specifications

Boiling Point 48°C to 49°C (0.02 mmHg)
Quantity 5 g
Solubility Information Soluble in dichloromethane,ethyl acetate.
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material Ethyl cyclohexylideneacetate
EINECSNumber 216-297-7
TSCA No

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.