Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl β-D-glucopyranoside | 3198-49-0 | 25 MG
SDP

Catalog No. 5000421803
Encompass_Preferred

Ethyl β-D-glucopyranoside is an ethyl derivative that can be isolated from the ethanolic extract of Scabiosa stellata LS.

  • Store at 4°C, protect from light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protect from light.
  • Soluble in DMSO at 100 mg/mL.
  • Suitable for in vivo experiments with various solvent systems, achieving solubility of ≥ 2.5 mg/mL.

Catalog No. 50-004-21803 Supplier Medchemexpress LLC Supplier No. 25MGHYN3872
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl β-D-glucopyranoside is an ethyl derivative that can be isolated from the ethanolic extract of Scabiosa stellata LS.

  • Store at 4°C, protect from light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protect from light.
  • Soluble in DMSO at 100 mg/mL.
  • Suitable for in vivo experiments with various solvent systems, achieving solubility of ≥ 2.5 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.