Learn More
Medchemexpress LLC Ethyl β-D-glucopyranoside | 3198-49-0 | 5 MG
SDP
Catalog No. 5000420967
Encompass_Preferred
Ethyl β-D-glucopyranoside (compound 10) is an ethyl derivative that can be isolated from the ethanolic extract of Scabiosa stellata LS. It is intended for research use only.
  • Store at 4°C, protected from light.
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
Catalog No. 50-004-20967 Supplier Medchemexpress LLC Supplier No. HYN38725MG
Ethyl β-D-glucopyranoside (compound 10) is an ethyl derivative that can be isolated from the ethanolic extract of Scabiosa stellata LS. It is intended for research use only.
  • Store at 4°C, protected from light.
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.