Learn More
Medchemexpress LLC Ethyl-α-D-thioglucopyranoside | 13533-58-9 | 97.0% | 224.28 | C8H16O5S | 250 MG
SDP
Catalog No. 5000303006
Encompass_Preferred
Ethyl-α-D-thioglucopyranoside is a thio-glycoside reagent used as a glycosyl donor in carbohydrate chemistry and biochemical assays. Supplied as a white to off-white solid, it has molecular formula C8H16O5S, molecular weight 224.28, and a reported purity of 97.0% (NMR). Recommended storage conditions are powder at -20°C (3 years) or 4°C (2 years); in solution store at -80°C (6 months) or -20°C (1 month).
  • Used as a glycosyl donor in carbohydrate synthesis and enzymatic studies.
  • High purity confirmed by NMR (97.0%).
  • White to off-white solid, easy to handle and store.
  • Powder stability: -20°C (3 years), 4°C (2 years).
  • Solution stability: -80°C (6 months), -20°C (1 month).
  • Molecular formula C8H16O5S; molecular weight 224.28.
Catalog No. 50-003-03006 Supplier Medchemexpress LLC Supplier No. 250MGHYW039849
Ethyl-α-D-thioglucopyranoside is a thio-glycoside reagent used as a glycosyl donor in carbohydrate chemistry and biochemical assays. Supplied as a white to off-white solid, it has molecular formula C8H16O5S, molecular weight 224.28, and a reported purity of 97.0% (NMR). Recommended storage conditions are powder at -20°C (3 years) or 4°C (2 years); in solution store at -80°C (6 months) or -20°C (1 month).
  • Used as a glycosyl donor in carbohydrate synthesis and enzymatic studies.
  • High purity confirmed by NMR (97.0%).
  • White to off-white solid, easy to handle and store.
  • Powder stability: -20°C (3 years), 4°C (2 years).
  • Solution stability: -80°C (6 months), -20°C (1 month).
  • Molecular formula C8H16O5S; molecular weight 224.28.