Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl diazoacetate solution

Catalog No. 5000031038 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-000-31038 25 mL
75-215-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-000-31038 Supplier Sigma Aldrich Supplier No. 75215025ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

New Diazoacetate Formulations

Specifications

CAS 623-73-4
Density 0.895 g/mL (at 25°C)
Molecular Formula C4H6N2O2
Linear Formula C4H6N2O2
Refractive Index n20/D 1.505
MDL Number MFCD00001989
Quantity 25 mL
Synonym 2-Diazoacetic acid ethyl ester solution
Molecular Weight (g/mol) 114.1
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.