Learn More
Medchemexpress LLC Ethyl docos-13-enoat 25g
SDP
Catalog No. 5000206133
Encompass_Preferred
Ethyl docos-13-enoate is a colorless or pale yellow liquid with a sweet floral aroma used as a flavoring agent in the food industry and as a flavor enhancer in perfumes and colognes It is also found in some vegetable oils and is used as a surfactant and emulsifier in personal care products Ethyl docos-13-enoate is a versatile compound used in various industries due to its pleasant odor and ability to mix ingredients together
Catalog No. 50-002-06133 Supplier Medchemexpress LLC Supplier No. 25GHYW099644
Ethyl docos-13-enoate is a colorless or pale yellow liquid with a sweet floral aroma used as a flavoring agent in the food industry and as a flavor enhancer in perfumes and colognes It is also found in some vegetable oils and is used as a surfactant and emulsifier in personal care products Ethyl docos-13-enoate is a versatile compound used in various industries due to its pleasant odor and ability to mix ingredients together