Learn More
Medchemexpress LLC HY-W011120 50mg Medchemexpress, Ethyl docosa-4,7,10,13,16,19-hexaenoate CAS:84494-72-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01112050MG
Encompass_Preferred
Medchemexpress, HY-W011120 50mg Ethyl docosa-4,7,10,13,16,19-hexaenoate CAS:84494-72-4 Ethyl cis-4,7,10,13,16,19-Docosahexaenoate, the ethyl ester of Docosahexaenoate (DHA), is enriched in the ethyl ester fraction by the selective alcoholysis of fatty acid ethyl esters originating from tuna oil with lauryl alcohol. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4548