Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl (E)-2-hexenoate | 27829-72-7 | MFCD00036541 | 98.5% | 142.20 g/mol | C8H14O2 | 10 G
SDP

Catalog No. 5000300622
Encompass_Preferred

Ethyl (E)-2-hexenoate is an organic ester used as a research chemical and reference compound in flavor, fragrance, and synthetic chemistry. It is supplied as a liquid with high purity and well-documented storage and safety data; consult the SDS for handling and transport details.

  • High purity suitable for analytical and research use.
  • Liquid form with density approximately 0.95 g/cm³.
  • Molecular formula C8H14O2; molecular weight 142.20 g/mol.
  • Commonly used as a reference in flavor and fragrance research and in synthesis.
  • Storage and handling guidance available in the SDS; follow recommended temperature controls.

Catalog No. 50-003-00622 Supplier Medchemexpress LLC Supplier No. HYW01756710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl (E)-2-hexenoate is an organic ester used as a research chemical and reference compound in flavor, fragrance, and synthetic chemistry. It is supplied as a liquid with high purity and well-documented storage and safety data; consult the SDS for handling and transport details.

  • High purity suitable for analytical and research use.
  • Liquid form with density approximately 0.95 g/cm³.
  • Molecular formula C8H14O2; molecular weight 142.20 g/mol.
  • Commonly used as a reference in flavor and fragrance research and in synthesis.
  • Storage and handling guidance available in the SDS; follow recommended temperature controls.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.