Learn More
Medchemexpress LLC Ethyl (E)-p-hydroxycinnamate | 7362-39-2 | MFCD00210460 | 99.2% | 192.21 g/mol | C11H12O3 | 10 MG
SDP
Catalog No. 5000289652
Encompass_Preferred
p-Coumaric Acid Ethyl Ester is the ethyl ester of p-coumaric acid, used in biochemical research as a reversible, non-competitive inhibitor of tyrosinase and as a building block in studies of enzyme inhibition, formulation, and cosmetic or food preservation research.
  • Reversible, non-competitive tyrosinase inhibitor (activity reported by manufacturer).
  • High purity: 99.19% (manufacturer reported).
  • Molecular weight 192.21 g/mol and formula C11H12O3.
  • Available as solid (5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and as a 10 mM solution in DMSO for ready-to-use assays.
  • Suitable for medicinal, cosmetic, and fruit preservation research applications.
Catalog No. 50-002-89652 Supplier Medchemexpress LLC Supplier No. HYN310310MG
p-Coumaric Acid Ethyl Ester is the ethyl ester of p-coumaric acid, used in biochemical research as a reversible, non-competitive inhibitor of tyrosinase and as a building block in studies of enzyme inhibition, formulation, and cosmetic or food preservation research.
  • Reversible, non-competitive tyrosinase inhibitor (activity reported by manufacturer).
  • High purity: 99.19% (manufacturer reported).
  • Molecular weight 192.21 g/mol and formula C11H12O3.
  • Available as solid (5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and as a 10 mM solution in DMSO for ready-to-use assays.
  • Suitable for medicinal, cosmetic, and fruit preservation research applications.