Learn More
eMolecules​ 87-13-8 | Ethyl ethoxymethylene malonate | Oakwood Chemical | MFCD00009148 | 216.233 | C10H16O5 | 98.000 | CCOC=C(C(=O)OCC)C(=O)OCC | 500g | 537687906
Catalog No. NC2221170 Shop All eMolecules Inc. Products
Encompass_Preferred
Ethyl ethoxymethylene malonate | Oakwood Chemical | 87-13-8 | MFCD00009148 | 216.233 | C10H16O5 | 98.000 | CCOC=C(C(=O)OCC)C(=O)OCC | 500g | 537687906
This item is not returnable. View return policy
Catalog No. NC2221170 Supplier eMolecules​ Supplier No. 537687906
Ethyl ethoxymethylene malonate | Oakwood Chemical | 87-13-8 | MFCD00009148 | 216.233 | C10H16O5 | 98.000 | CCOC=C(C(=O)OCC)C(=O)OCC | 500g | 537687906