Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (ethoxymethylene)cyanoacetate, 98%

Catalog No. p-128402
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118481000 100 g Glass bottle
AC118485000 500 g Glass bottle
AC118480010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC118481000 Supplier Thermo Scientific Chemicals Supplier No. 118481000
Only null left
Add to Cart

CAS: 94-05-3 | C8H11NO3 | 169.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-05-3
Molecular Formula C8H11NO3
Molecular Weight (g/mol) 169.18
MDL Number MFCD00009136
InChI Key KTMGNAIGXYODKQ-SREVYHEPSA-N
Synonym ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoate
PubChem CID 1715183
SMILES CCO\C=C(\C#N)C(=O)OCC

Specifications

Melting Point 48°C to 52°C
Color White to Yellow
Boiling Point 190°C to 191°C (30.0 mmHg)
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OCH=C(CN)CO2C2H5
Quantity 100 g
Beilstein 03,47
Fieser 01,38
Solubility Information Solubility in water: <0.01g/L (20°C)
IUPAC Name ethyl (2Z)-2-cyano-3-ethoxyprop-2-enoate
Formula Weight 169.18
Percent Purity 98%
Physical Form Crystalline Mass or Crystals
Chemical Name or Material Ethyl (ethoxymethylene)cyanoacetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Causes
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mis
EINECSNumber 202-299-5
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.