Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl ethynyl ether, 50 wt.% solution in hexanes, AcroSeal™

Catalog No. AC429061000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429061000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC429061000 Supplier Thermo Scientific Chemicals Supplier No. 429061000
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 927-80-0 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl ethynyl ether
CAS 927-80-0, 92112-69-1
Color Orange to Brown
Density 0.7180g/mL
Boiling Point 55.0°C to 56.0°C
Flash Point −28°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C4H6O
Linear Formula C2H5OC≡CH
Quantity 100 mL
MDL Number MFCD00009247
Specific Gravity 0.718
Synonym ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
InChI Key WMYNMYVRWWCRPS-UHFFFAOYSA-N
SMILES CCOC#C
IUPAC Name ethynoxyethane
Molecular Weight (g/mol) 70.09
PubChem CID 61239
Formula Weight 70.09
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.