Learn More
Apexbio Technology LLC ETHYL-FERULATE-20MG
SDP
Catalog No. 5000570709
Encompass_Preferred
Ethyl ferulate (CAS No 4046-02-0) is a compound studied in biomedical research for its antioxidant and anti-inflammatory properties Originating from the esterification of ferulic acid it engages in mechanisms involving the inhibition of oxidative stress pathways and modulation of inflammatory cytokines In vitro its activity is typically observed in the nanomolar to low micromolar range Common applications include investigating its effects in various cell models such as fibroblasts and macrophages and in animal studies for evaluating potential therapeutic effects against chronic diseases It is also utilized in drug screening processes to explore new pharmaceutical applications Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70709 Supplier Apexbio Technology LLC Supplier No. N244420
Ethyl ferulate (CAS No 4046-02-0) is a compound studied in biomedical research for its antioxidant and anti-inflammatory properties Originating from the esterification of ferulic acid it engages in mechanisms involving the inhibition of oxidative stress pathways and modulation of inflammatory cytokines In vitro its activity is typically observed in the nanomolar to low micromolar range Common applications include investigating its effects in various cell models such as fibroblasts and macrophages and in animal studies for evaluating potential therapeutic effects against chronic diseases It is also utilized in drug screening processes to explore new pharmaceutical applications Experimental concentrations typically depend on the specific research design