Learn More
Selleck Chemical LLC Ethyl ferulate S3855-25mg
SDP
Catalog No. 503109076
Encompass_Preferred
Ethyl ferulate (Ferulic acid ethyl ester) is the alkyl ester derivative of ferulic acid which is a naturally occurring plant product with anti-oxidative anti-inflammatory neuroprotective and antiproliferative activities
Catalog No. 50-310-9076 Supplier Selleck Chemical LLC Supplier No. S385525MG
Ethyl ferulate (Ferulic acid ethyl ester) is the alkyl ester derivative of ferulic acid which is a naturally occurring plant product with anti-oxidative anti-inflammatory neuroprotective and antiproliferative activities