Learn More
Medchemexpress LLC Ethyl ferulate | 4046-02-0 | 99.9% | 222.24 | 1 ML
SDP
Catalog No. 5000427977
Encompass_Preferred
Ethyl ferulate is a naturally lipophilic derivative of ferulic acid, originally derived from Rhizoma Chuanxiong. It induces heme oxygenase-1 (HO-1) and offers neuroprotection against oxidative stress and amyloid β peptide (1-42)-induced neurotoxicity.
  • Induces heme oxygenase-1 (HO-1) expression.
  • Protects neurons from oxidative stress.
  • Reduces amyloid β peptide (1-42)-induced neurotoxicity.
  • Increases HO activity in astrocytes and hippocampal neurons.
  • Inhibits LPS-induced acute lung injury.
  • Inhibits retinal neovascularization.
Catalog No. 50-004-27977 Supplier Medchemexpress LLC Supplier No. HYN006110MM/1ML
Ethyl ferulate is a naturally lipophilic derivative of ferulic acid, originally derived from Rhizoma Chuanxiong. It induces heme oxygenase-1 (HO-1) and offers neuroprotection against oxidative stress and amyloid β peptide (1-42)-induced neurotoxicity.
  • Induces heme oxygenase-1 (HO-1) expression.
  • Protects neurons from oxidative stress.
  • Reduces amyloid β peptide (1-42)-induced neurotoxicity.
  • Increases HO activity in astrocytes and hippocampal neurons.
  • Inhibits LPS-induced acute lung injury.
  • Inhibits retinal neovascularization.