Learn More
Medchemexpress LLC Ethyl ferulate | 4046-02-0 | 99.9% | 222.24 | 5 G
SDP
Catalog No. 5000427978
Encompass_Preferred
Ethyl ferulate is a naturally occurring lipophilic derivative of ferulic acid, originally found in Rhizoma Chuanxiong. It induces heme oxygenase-1 (HO-1) and protects rat neurons against oxidative stress. It also protects neurons from amyloid β peptide (1-42)-induced oxidative stress and neurotoxicity.
  • Induces heme oxygenase-1 (HO-1)
  • Protects rat neurons against oxidative stress
  • Protects neurons against amyloid β peptide (1-42)-induced oxidative stress and neurotoxicity
  • Inhibits LPS-induced acute lung injury in mice
  • Inhibits retinal neovascularization in oxygen-induced retinopathy mice model
  • Increases Nrf-2 expression and nuclear translocation
  • Decreases Keap-1 expression, and increases HO-1 and NQO-1 expression
  • Reduces hypoxia-induced HIF-1α and VEGFA expression
Catalog No. 50-004-27978 Supplier Medchemexpress LLC Supplier No. HYN00615G
Ethyl ferulate is a naturally occurring lipophilic derivative of ferulic acid, originally found in Rhizoma Chuanxiong. It induces heme oxygenase-1 (HO-1) and protects rat neurons against oxidative stress. It also protects neurons from amyloid β peptide (1-42)-induced oxidative stress and neurotoxicity.
  • Induces heme oxygenase-1 (HO-1)
  • Protects rat neurons against oxidative stress
  • Protects neurons against amyloid β peptide (1-42)-induced oxidative stress and neurotoxicity
  • Inhibits LPS-induced acute lung injury in mice
  • Inhibits retinal neovascularization in oxygen-induced retinopathy mice model
  • Increases Nrf-2 expression and nuclear translocation
  • Decreases Keap-1 expression, and increases HO-1 and NQO-1 expression
  • Reduces hypoxia-induced HIF-1α and VEGFA expression