Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl fluoroacetate

Catalog No. 16381310G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-381-310G 10 g
16-381-350G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-381-310G Supplier Sigma Aldrich Supplier No. 16381310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl fluoroacetate is an ester organic solvent.ethyl fluoroacetate was used in the synthesis of 4-deoxy-4-fluoro-muscarines
  • It was used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate
  • It was used in development of an ammonolysis-based microencapsulation process

Specifications

CAS 459-72-3
Density 1.098 g/mL (at 25°C (literature))
Boiling Point 119.3°C (753 mmHg)
Molecular Formula C4H7FO2
Refractive Index n20/D 1.375 (literature)
Linear Formula FCH2COOC2H5
MDL Number MFCD00000450
Quantity 10 g
Molecular Weight (g/mol) 106.1
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.