Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl Formate

Catalog No. 11268225L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-268-225L 2.5 L
11-268-225ML 25 mL
11-268-2500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-268-225L Supplier Sigma Aldrich Supplier No. 1126822.5L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl Formate

Specifications

CAS 109-94-4
Melting Point -80°C
Density 0.921 g/mL at 20°C
Boiling Point 52°C to 54°C
Molecular Formula C3H6O2
Refractive Index n20/D 1.359
Linear Formula HCOOC2H5
MDL Number MFCD00003294
Quantity 2.5 L
Synonym Formic acid ethyl ester
Molecular Weight (g/mol) 74.08 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.