Learn More
Medchemexpress LLC Ethyl gallate | 831-61-8 | 198.17 | 1 G
SDP
Catalog No. 5000389870
Encompass_Preferred
Ethyl gallate is a nonflavonoid phenolic compound that acts as a scavenger of hydrogen peroxide. It exhibits cytotoxic effects on HL-60 cells, inducing apoptosis, decreasing cell viability in a time- and dose-dependent manner, and increasing cells in the subG1 phase. It also modulates protein expression by decreasing Bcl-2 and increasing Bax and truncated Bid (tBid).
  • Nonflavonoid phenolic compound
  • Scavenges hydrogen peroxide
  • Induces apoptosis in HL-60 cells
  • Decreases cell viability in a time- and dose-dependent manner
  • Increases proportion of cells in subG1 phase
  • Modulates protein expression by decreasing Bcl-2 and increasing Bax and truncated Bid (tBid)
Catalog No. 50-003-89870 Supplier Medchemexpress LLC Supplier No. HYN05251G
Ethyl gallate is a nonflavonoid phenolic compound that acts as a scavenger of hydrogen peroxide. It exhibits cytotoxic effects on HL-60 cells, inducing apoptosis, decreasing cell viability in a time- and dose-dependent manner, and increasing cells in the subG1 phase. It also modulates protein expression by decreasing Bcl-2 and increasing Bax and truncated Bid (tBid).
  • Nonflavonoid phenolic compound
  • Scavenges hydrogen peroxide
  • Induces apoptosis in HL-60 cells
  • Decreases cell viability in a time- and dose-dependent manner
  • Increases proportion of cells in subG1 phase
  • Modulates protein expression by decreasing Bcl-2 and increasing Bax and truncated Bid (tBid)