Learn More
TARGETMOL CHEMICALS INC Ethyl glucoside 50MG
SDP
Catalog No. 502875835
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Ethyl glucoside (beta-D-Glucopyranoside, ethyl), a shale inhibitor for water-based drilling fluids, is a versatile initiator of enzyme-catalyzed regioselective lactone ring-opening polymerization and is a natural compound found in Sisyrinchium palmifolium. Purity 99.68%
Catalog No. 50-287-5835 Supplier TARGETMOL CHEMICALS INC Supplier No. TN240150MG
Also available in 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Ethyl glucoside (beta-D-Glucopyranoside, ethyl), a shale inhibitor for water-based drilling fluids, is a versatile initiator of enzyme-catalyzed regioselective lactone ring-opening polymerization and is a natural compound found in Sisyrinchium palmifolium. Purity 99.68%