Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl glucuronide | 17685-04-0 | 99.2% | 222.19 | 1 ML
SDP

Catalog No. 5000435603
Encompass_Preferred

Ethyl glucuronide is an endogenous metabolite of ethanol. It serves as a biomarker for ethanol exposure, accumulating in hair and reflecting alcohol intake over time. It is also an agonist for TLR4.

  • Endogenous metabolite
  • Biomarker for ethanol exposure
  • Agonist for TLR4
  • Appearance: Solid, white to off-white
  • Storage: -20°C, protect from light, stored under nitrogen. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light, stored under nitrogen).
  • Useful in research for studying ethanol exposure and its effects
  • Can be used to block the formation of the TLR4-MD2 complex and stimulate the expression of TLR4-dependent SEAP protein in HEK-TLR4 cells
  • Has been shown to reduce tactile threshold and induce hyperalgesia in Sprague-Dawley rat models

Catalog No. 50-004-35603 Supplier Medchemexpress LLC Supplier No. HY11309310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl glucuronide is an endogenous metabolite of ethanol. It serves as a biomarker for ethanol exposure, accumulating in hair and reflecting alcohol intake over time. It is also an agonist for TLR4.

  • Endogenous metabolite
  • Biomarker for ethanol exposure
  • Agonist for TLR4
  • Appearance: Solid, white to off-white
  • Storage: -20°C, protect from light, stored under nitrogen. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light, stored under nitrogen).
  • Useful in research for studying ethanol exposure and its effects
  • Can be used to block the formation of the TLR4-MD2 complex and stimulate the expression of TLR4-dependent SEAP protein in HEK-TLR4 cells
  • Has been shown to reduce tactile threshold and induce hyperalgesia in Sprague-Dawley rat models

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.