Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl glutamate | 1119-33-1 | MFCD00037217 | 97.0% | 175.18 | C7H13NO4 | 50G
SDP

Catalog No. 5000236890
Encompass_Preferred

Ethyl glutamate is the ethyl ester derivative of glutamic acid supplied as a white to off-white solid for life-science research. It is described as promoting cell growth and is provided with supporting documentation including a datasheet, safety data sheet, and certificate of analysis. Typical purity is 97.0%.

  • CAS number 1119-33-1.
  • Purity 97.0%.
  • Molecular formula C7H13NO4.
  • Molecular weight 175.18.
  • Appearance solid, white to off-white.
  • Package sizes available: 25 G, 50 G, 100 G, 500 G, bulk on request.
  • Storage store at room temperature; in solvent: -80°C (2 years), -20°C (1 year).
  • Applications used in life-science research and studies of cell growth.

Catalog No. 50-002-36890 Supplier Medchemexpress LLC Supplier No. HY12422850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl glutamate is the ethyl ester derivative of glutamic acid supplied as a white to off-white solid for life-science research. It is described as promoting cell growth and is provided with supporting documentation including a datasheet, safety data sheet, and certificate of analysis. Typical purity is 97.0%.

  • CAS number 1119-33-1.
  • Purity 97.0%.
  • Molecular formula C7H13NO4.
  • Molecular weight 175.18.
  • Appearance solid, white to off-white.
  • Package sizes available: 25 G, 50 G, 100 G, 500 G, bulk on request.
  • Storage store at room temperature; in solvent: -80°C (2 years), -20°C (1 year).
  • Applications used in life-science research and studies of cell growth.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.